This product is now discontinued, View Related Products

TEDIA® Xylenes, ACS Reagent Grade

High-purity solvents for life science, biotechnology, pharmaceutical, laboratory, and industrial applications.
  • Quality and consistency your lab can count on
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Tedia Xylenes, ACS Reagent Grade, 4 x 4 L EW-15001-98 Mfr # XR2250-001
COMPARE
ACS Reagent 16000 Xylenes, ACS Reagent Grade, 4 x 4 L
Call For Price
Tedia Xylenes, ACS Reagent Grade; 6 x 1 L, Amber Glass EW-15001-99 Mfr # XR2250-002
COMPARE
ACS Reagent 6000 Xylenes, ACS Reagent Grade; 6 x 1 L, Amber Glass
Call For Price
Tedia Xylenes, Histological Grade; 4 x 4 L, Amber Glass EW-15002-01 Mfr # XR2251-001
COMPARE
Histological 16000 Xylenes, Histological Grade; 4 x 4 L, Amber Glass
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat