This product is now discontinued, View Related Products
Tefzel Adapters
Connect your micro tubing to standard size tubing or luer fitting systems. These TEFZEL® adapters are designed for use with small tubing fittings. Adapters come with either caps or with a 1/4-28 UNF thread and lock nut. Adapters with caps include zero dead volume PTFE cones and screw onto the fitting body. Adapters with 1/4-28 UNF thread come with lock nuts and screw directly into existing fitting caps.
$20.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Tefzel Adapters with caps; 1/4" OD pipe; EW-06471-82 Mfr # 001203
COMPARE
Adapters with caps; 1/4\" OD pipe;
$20.00USD / Each
Tefzel Adapters with caps; 1/8" OD pipe; EW-06471-83 Mfr # 001204
COMPARE
Adapters with caps; 1/8\" OD pipe;
$20.00USD / Each
Tefzel adapters, Adapter with caps, 1/16" OD tubing, 316SS EW-06471-84 Mfr # 001205
COMPARE
adapters, Adapter with caps, 1/16\" OD tubing, 316SS
Call For Price
Tefzel ETFE Adapter with 1/4-28 UNF Thread, 1/4 EW-06471-92 Mfr # 002503
COMPARE
ETFE Adapter with 1/4-28 UNF Thread, 1/4
Call For Price
Tefzel ETFE Adapter with 1/4-28 UNF Thread, 1/8 EW-06471-93 Mfr # 002504
COMPARE
ETFE Adapter with 1/4-28 UNF Thread, 1/8
Call For Price
Tefzel ETFE Adapter with 1/4-28 UNF Thread, 1/16 EW-06471-94 Mfr # 002505
COMPARE
ETFE Adapter with 1/4-28 UNF Thread, 1/16