Cole-Parmer® Temperature Compensated Barometers
Monitor barometric pressure with these wall-mount barometers
  • Ideal for monitoring impending weather changes
  • Barometers are encased in brass, with bezel for bulkhead mounting
  • Measure 2" depth x 6.1" diameter
Cole-Parmer Temperature Compensated Barometers measure atmospheric pressure in millibar (mbar) and inches Hg or millibar and mm Hg. Barometers also monitor temperature and provide temperature compensation with a built-in bi-metal thermometer.
ITEM(S) ON SALE
$405.20 - $424.40USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Barometric Pressure Resolution (PSI) Max Temperature (Outside) (° F) Temperature Accuracy (Outside) (° F)
Availability Pricing
COMPARE
1 mbar (0.1\" Hg) 122 ±1.8
$424.40USD / Each
$530.50USD / Each
COMPARE
1 mbar (1 mm Hg) 122 ±1.8
$405.20USD / Each
$506.50USD / Each