This product is now discontinued, View Related Products
Ten-Gallon Heavy-Duty Ultrasonic Cleaner
Continuous duty design is ideal for your rigorous cleaning needs
Use this cleaner with mildly acidic, neutral, or alkaline aqueous solutions to clean large glass, plastic, or metal objects. High-efficiency, built-in frequency generator with RFI suppression is designed for continuous duty. Built-in, 1000-watt heater maintains solution temperature from ambient to 140°F (60°C). Operates at 40 kHz.
Easy-to-clean, deep-drawn 304 stainless steel tank has no corner welds or other dirt-catching surfaces; drain is bottom-mounted. Corrosion-resistant tank is insulated for quiet operation and low heat loss.
Note: Always suspend parts to be cleaned with a wire or use perforated accessory basket 18000-72 sold separately.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Branson Ten-gallon heavy-duty ultrasonic cleaner, 120 VAC EW-08847-00 Mfr # 000-914-506
COMPARE
120
Call For Price
Branson Ten-gallon heavy-duty ultrasonic cleaner, 220 VAC EW-08847-02 Mfr # 000-914-606
COMPARE
220