Test Products Digital Manometers
Replace messy U-tubes (water column manometers) and expensive pressure gauges with digital manometers.
FEATURES
  • Measure pressure of air and natural gas
  • Auto Power Off after 40 minutes
      APPLICATIONS
      • Adjusting appliance regulators
      • Setting delivery pressures
      • Leakage "shut-in" test
      • Regulator adjustments for pressure and lock-up tests
      • Test for leaks in low-pressure gas systems
      • Isolate difficult pressure problems on natural gas and propane systems
      • Test pressure switch operation on a 2-stage gas furnace in both positive and negative pressures
      • Static pressures in air ducts for air balancing
      • Check air filters by measuring pressure drop
$150.00 - $200.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Pressure (in WC) Description
Availability Pricing
COMPARE
±0.5% of reading 5 digits 120 Dual Input Differential Manometer, /-5 psi
$200.00USD / Each
COMPARE
±0.5% of reading 5 digits 48 Manometer /-80.29 in. H2O, Single Input
$150.00USD / Each
COMPARE
±0.5% of reading 5 digits 48 Digital Manometer 15/48in.H2O Single Input, 7 Selectable Units of Measure
$193.00USD / Each