Testo Multi-Function Meter - Logger for HVAC and IAQ
Use anywhere you need to take important IAQ measurements
  • Large backlit display
  • Simple menu operation
  • Programmed functions such as mean calculations and area inputs for volume flow calculations
  • IP54 rating for meter
  • Rubberized case for shock and impact protection
  • Integrated magnets on the meter for hands-free operation
The Testo 435 is perfect for monitoring, analyzing, and diagnosing indoor air quality problems in the office, warehouse, on the production floor, or anywhere you need to take important IAQ measurements.
$91.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Testo 0554 0190 Wireless Upgrade Module for Model 625 Thermohygrometer EW-37852-53 Mfr # 0554 0190
COMPARE
Wireless Upgrade Module for Model 625 Thermohygrometer
$91.00USD / Each