This product is now discontinued, View Related Products
Thermo Scientific™ ART™ Pipette Tips, Non-Sterile
No cross-contamination by aerosols and liquids
  • Self-sealing barrier eliminates cross-contamination, even when samples are overdrawn
  • Tips load and eject easily for reduced hand fatigue
  • Certified RNase-, DNase, and pyrogen free—ideal for use in genetic studies, forensics, PCR, and radioisotope sampling
The Aerosol-Resistant Tip (ART™) is a self-sealing filter tip that protects your pipettor from crossover contamination by eliminating contact with liquids or aerosols. Tips are made of polypropylene. Special options include low retention to minimize sample loss, Micropoint™ design for precise control, extended length for
contamination-free reach into vessels, and gel loading for electrophoresis preparation.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Max Volume (µL) Overall Length (in)
Availability Pricing
Thermo Scientific Art Tips 2139B Pipette Tips, 0.1-10 µL, MT, 1000/bag; Finn EW-25716-02 Mfr # 2139B
COMPARE
10 1.25
Call For Price
Thermo Scientific Art Tips 2140B Pipette Tips, 0.1-10 µL, MT/E x L, 1000/bag; Finn EW-25716-08 Mfr # 2140B
COMPARE
10 1.75
Call For Price
Thermo Scientific Art Tips 2149EB Pipette Tips, 0.1-10 µL, 1000/bag; Epp EW-25716-12 Mfr # 2149EB
COMPARE
10 1.875
Call For Price
Thermo Scientific Art Tips 2149PB Pipette Tip, 0.5-20µL, 1000/pk EW-25716-22 Mfr # 2149PB
COMPARE
20 2
Call For Price
Thermo Scientific Art Tips 2155B Tips Pipette Tip, 0.5-100µL, 1000/pk EW-25716-28 Mfr # 2155B
COMPARE
100 2.75
Call For Price
Thermo Scientific Art Tips 2065EB Pipette Tip, 1-100µL, 1000/pk EW-25716-32 Mfr # 2065EB
COMPARE
100 2
Call For Price
Thermo Scientific Art Tips 2159PB Pipette Tip, 5-180µL, 1000/pk EW-25716-36 Mfr # 2159PB
COMPARE
180 3.375
Call For Price
Thermo Scientific Art Tips 2069B Pipette Tip, 1-200µL, 1000/pk EW-25716-40 Mfr # 2069B
COMPARE
200 2.6
Call For Price
Thermo Scientific Art Tips 2160PB Tips Pipette Tip, 5-200µL, 1000/pk EW-25716-44 Mfr # 2160PB
COMPARE
200 3.375
Call For Price
Thermo Scientific Art Tips 2070B Pipette Tip, 1-300µL, 1000/pk EW-25716-48 Mfr # 2070B
COMPARE
300 2.375
Call For Price
Thermo Scientific Art Tips 2279B Pipette Tip, 100-1000µL, 1000/pk EW-25716-52 Mfr # 2279B
COMPARE
1000 3.125
Call For Price
Thermo Scientific Art Tips 2279EB Tips Pipette Tip, 100-1000µL, 1000/pk EW-25716-54 Mfr # 2279EB
COMPARE
1000 3.125
Call For Price
Showing 1- 12 of 12