Thermo Scientific™ ART™ Pipette Tips, Sterile
Racked and presterilized pipette tips
  • Self-sealing barrier eliminates cross-contamination, even when samples are overdrawn
  • Tips load and eject easily for reduced hand fatigue
  • Certified RNase-, DNase, and pyrogen free—ideal for use in genetic studies, forensics, PCR, and radioisotope sampling
The Aerosol-Resistant Tip (ART™) is a self-sealing filter tip that protects your pipettor from crossover contamination by eliminating contact with liquids or aerosols. Tips are made of polypropylene. Special options include low retention to minimize sample loss, Micropoint™ design for precise control, extended length for
contamination-free reach into vessels, and gel loading for electrophoresis preparation.
$218.00 - $268.00USD / Pkg of 1
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Overall Length (in) Description
Availability Pricing
Thermo Scientific Art Tips 2139 Pipette Tips, 0.1-10 µL, Sterile, MT, 10 Trays x 96; Finn EW-25716-04 Mfr # 2139
COMPARE
10 1.25 Standard Retention
$220.00USD / Pkg of 1
Thermo Scientific Art Tips 2140-05 Pipette Tips, 0.1-10 µL, Sterile, LR, MT, 10 Trays x 96; Finn EW-25716-06 Mfr # 2140-05
COMPARE
10 1.75 Low Retention
$245.00USD / Pkg of 1
Thermo Scientific Art Tips 2149E Pipette Tips, 0.1-10 µL, Sterile, 10 Trays x 96; Epp EW-25716-14 Mfr # 2149E
COMPARE
10 1.875 Standard Retention
$218.00USD / Pkg of 1
Thermo Scientific Art Tips 2149 Pipette Tips, 0.1-10 µL, Sterile, MT, 10 Trays x 96; Epp EW-25716-16 Mfr # 2149
COMPARE
20 1.875 Standard Retention
$222.00USD / Pkg of 1
Thermo Scientific Art Tips 2149P-05 Pipette Tips, 0.5-20 µL, Sterile, LR, 10 Trays x 96; Pman EW-25716-18 Mfr # 2149P-05
COMPARE
20 2 Low Retention
$245.00USD / Pkg of 1
Thermo Scientific Art Tips 2149P Pipette Tips, 0.5-20 µL, Sterile, MT, 10 Trays x 96; Pman EW-25716-20 Mfr # 2149P
COMPARE
20 2 Standard Retention
$223.00USD / Pkg of 1
Thermo Scientific Art Tips 2155P Pipette Tips, 0.5-20uL, Sterile, MT/GL, 10 traysx96; Finn EW-25716-24 Mfr # 2155P
COMPARE
20 2.75 Gel Loading
$268.00USD / Pkg of 1
Thermo Scientific Art Tips 2155 Pipette Tip, 0.5-100µL, Sterile, 960/pk EW-25716-26 Mfr # 2155
COMPARE
100 2.75 Gel Loading
$268.00USD / Pkg of 960
Thermo Scientific Art Tips 2065E Pipette Tip, 1-100µL, Sterile, 960/pk EW-25716-30 Mfr # 2065E
COMPARE
100 2 Standard Retention
$220.00USD / Pkg of 960
Thermo Scientific Art Tips 2069 Pipette Tip, 1-200µL, Sterile, 960/pk EW-25716-38 Mfr # 2069
COMPARE
200 2.375 Standard Retention
$226.00USD / Pkg of 960
Thermo Scientific Art Tips 2160P Pipette Tip, 5-200µL, Sterile, 960/pk EW-25716-42 Mfr # 2160P
COMPARE
200 3.5 Extended Length
$263.00USD / Pkg of 960
Thermo Scientific Art Tips 2279 Pipette Tip, 100-1000µL, Sterile, 768/pk EW-25716-50 Mfr # 2279
COMPARE
1000 3.125 Ergonomic
$220.00USD / Pkg of 8
Thermo Scientific Art Tips 2079E Pipette Tips, 100-1000uL, Sterile, 8 traysx100; Finn EW-25716-56 Mfr # 2079E
COMPARE
1000 3.5 Standard Retention
$222.00USD / Pkg of 1
Thermo Scientific Art Tips 2079 Pipette Tip, 100-1000µL, Sterile, 800/pk EW-25716-58 Mfr # 2079
COMPARE
1000 4.6 Extended Length
$238.00USD / Pkg of 800
Showing 1- 14 of 14