Thermo Scientific Barnstead Mega-Pure Glass Stills
High-purity water on demand
  • Nonmetal wetted parts ensure your water quality
These stills remove most dissolved solids and all pyrogens and biological impurities from tap water or pretreated water—meets pyrogen purity standards. Wetted parts are quartz, glass, and PTFE.
Stills feature a water-level sensor in the boiling chamber. The high-temperature cutoff switch prevents any possibility of overheating. Aluminum cabinet with baked-epoxy finish keeps electrical cords and the drain line out of your way.
Double solenoid valve lets you use pretreated feed water for feed to the boiler. This reduces the amount of cleaning required. Stills do not operate automatically with storage tanks other than the automatic collection system.
Order reservoir and valve separately.
System requires:
1) Mega-Pure glass still
2) Automatic collection system
3) Pretreatment dionizer (for systems with hard water makeup) and cartridges
$4,915.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Production (L/hour) Power (VAC)
Availability Pricing
Thermo Scientific Barnstead A440267 Glass Still, 1.0 LPH, 120 VAC EW-99292-00 Mfr # A440267
COMPARE
1 120
$4,915.00USD / Each