This product is now discontinued, View Related Products
Thermo Scientific Barnstead Pacfic RO Water Purfication Systems
Specifically developed for functional and economical production of high purity water
  • Integrated feed water monitoring ensures water quality and extends cartridge service life
  • Easy-to-operate clear display tilts for optimal viewing
  • Reservoir fill level (%) automatically shown on display
The Barnstead Pacific RO system delivers up to 240L/day of Type III (RO) water from a tap feed water supply. These units are available in five product water capacities - ideal for applications such as preparing and diluting buffers or supplying lab equipment like autoclaves, water baths, glassware washers, and incubators. Space-saving design allows for system installation on benchtop or for wall mounting.
Microprocessor control offers automatic operation, as well as real-time clock records system of errors and faults. The code-protected operating system prevents unauthorized changes to settings. Use the RS-232 interface for safe documentation of all measured values and faults with date and time in compliance with GLP guidelines; order external printer separately.
Opaque polyethylene reservoirs (required; order separately) allow complete draining and efficient cleaning and disinfection. Reservoirs are available with an optional recirculation pump which protects high purity water from bacterial growth during standstill.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Production (L/hour)
Availability Pricing
Thermo Scientific Barnstead 50132385 Pacific RO Water Purification System 3 L/hr EW-99289-00 Mfr # 50132385
COMPARE
3
Call For Price
Thermo Scientific Barnstead 50132386 Pacific RO Water Purification System 7 L/hr EW-99289-10 Mfr # 50132386
COMPARE
7
Call For Price
Thermo Scientific Barnstead 50132387 Pacific RO Water Purification System 12 L/hr EW-99289-20 Mfr # 50132387
COMPARE
12
Call For Price
Thermo Scientific Barnstead 50132388 Pacific RO Water Purification System 20 L/hr EW-99289-30 Mfr # 50132388
COMPARE
20
Call For Price
Thermo Scientific Barnstead 50132389 Pacific RO Water Purification System 40 L/hr EW-99289-40 Mfr # 50132389
COMPARE
40