Thermo Scientific Barnstead Pacific TII Water Purification System
Multiple capacities available to support your daily requirements
  • Microprocessor controlled for automatic operation with continuous monitoring of all critical parameters
  • Reservoir fill level (%) automatically shown on display
Systems deliver up to 240 L/day of Type II pure water from a tap feed water supply. Ideal for routine lab work such as reagent preparation, media preparation, and glassware rinsing. High performance reverse osmosis membrane removes approximately 98% of inorganic ions and 99% of all dissolved organic substances, as well as micoorganisms and particles.
Space-saving design allows benchtop operation or wall mounting. Large, illuminated four-line display is easy to read and clearly shows operating mode status (production, standby, disinfection), reservoir volume (%), and temperature (°C). Display can be tilted for optimal viewing.
Use the RS-232 interface for safe documentation of all measured values and faults with date and time in compliance with GLP guidelines; order external printer separately. Optional UV lamp (254 nm) acts as a germicide, preventing biological contamination.
Opaque polyethylene reservoirs (required; order separately) allow complete draining and efficient cleaning and disinfection. Reservoirs are available with an optional recirculation pump which protects high purity water from bacterial growth during standstill. The 30- and 60-L units can be used with the optional wall-mounting bracket for further space-saving capabilities.
$12,380.00 - $12,500.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Production (L/hour) With UV Lamp
Availability Pricing
Thermo Scientific Barnstead 50132125 Pacific TII Water Purification Systems 20 l/hr EW-99290-18 Mfr # 50132125
COMPARE
20 No
$12,500.00USD / Each
Thermo Scientific Barnstead 50132132 Pacific TII Water Purification Systems 12 l/hr UV EW-99290-26 Mfr # 50132132
COMPARE
12 Yes
$12,380.00USD / Each