This product is now discontinued, View Related Products

Thermo Scientific™ BioBasic SEC Columns

Provide superior separation of water soluble compounds
  • Ideally suited for sample clean-up
  • Analyte separation over a wide range of molecular weights
  • Long, efficient column life
BioBasic SEC (Size Exclusion Chromatography) columns are made with a mechanically rigid silica that does not swell or shrink with changes in solvent, providing higher efficiencies than polymer-based columns. The highly base-deactivated 5µm silica is polymer-coated to ensure separation only occurs on the basis of molecular size. Columns provide highly efficient separation of compounds with a molecular weight of 100 to 10,000,000.
Each lot of silica used in the production of BioBasic SEC columns is tested with a range of proteins to confirm accuracy. Each column is also tested for compliance with efficiency specifications and is shipped with a test certificate with both the column and silica efficiency data.
BioBasic SEC columns are ideal for gel filtration separation of proteins an
Call For Price

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific BioBasic 73305-037821 SEC 60 Guard Column - 30mm by 7.8mm, 60 Angstrom pore size EW-01902-07 Mfr # 73305-037821
COMPARE
Call For Price
Thermo Scientific BioBasic 73305-157846 SEC 60 Column - 150mm by 7.8mm, 60 Angstrom pore size EW-01902-08 Mfr # 73305-157846
COMPARE
7.8 150 0.006
Call For Price
Thermo Scientific BioBasic 73305-307846 SEC 60 Column - 300mm by 7.8mm, 60 Angstrom pore size EW-01902-09 Mfr # 73305-307846
COMPARE
Call For Price
Thermo Scientific BioBasic 73405-037821 SEC 120 Guard Column - 30mm by 7.8mm, 120 Angstrom pore size EW-01902-10 Mfr # 73405-037821
COMPARE
Call For Price
Thermo Scientific BioBasic 73405-157846 SEC 120 Column - 150mm by 7.8mm, 120 Angstrom pore size EW-01902-11 Mfr # 73405-157846
COMPARE
Call For Price
Thermo Scientific BioBasic 73405-307846 SEC 120 Column - 300mm by 7.8mm, 120 Angstrom pore size EW-01902-12 Mfr # 73405-307846
COMPARE
7.8 300 0.012
Call For Price
Thermo Scientific BioBasic 73505-037821 SEC 300 Guard Column - 30mm by 7.8mm, 300 Angstrom pore size EW-01902-13 Mfr # 73505-037821
COMPARE
Call For Price
Thermo Scientific BioBasic 73505-157846 SEC 300 Column - 150mm by 7.8mm, 300 Angstrom pore size EW-01902-14 Mfr # 73505-157846
COMPARE
Call For Price
Thermo Scientific BioBasic 73505-307846 SEC 300 Column - 300mm by 7.8mm, 300 Angstrom pore size EW-01902-15 Mfr # 73505-307846
COMPARE
7.8 300 0.03
Call For Price
Thermo Scientific BioBasic SEC 1000 Guard Column - 30mm by 7.8mm, 1000 Angstrom pore size EW-01902-16 Mfr # 73605-037821
COMPARE
7.8 30 0.1
Call For Price
Thermo Scientific BioBasic 73605-157846 SEC 1000 Column - 150mm by 7.8mm, 1000 Angstrom pore size EW-01902-17 Mfr # 73605-157846
COMPARE
Call For Price
Thermo Scientific BioBasic 73605-307846 SEC 1000 Guard Column - 300mm by 7.8mm, 1000 Angstrom pore size EW-01902-18 Mfr # 73605-307846
COMPARE
Call For Price
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat