This product is now discontinued, View Related Products
Thermo Scientific Flammable Storage and Explosion-Proof Refrigerators
Reduce the risk of explosion with a spark-free interior and exterior
  • Designed for use in Class I, Groups C & D & Class I, Zone 1, Group IIB hazardous environments
  • CFC-free insulation and refrigerant
These explosion proof refrigerators are particularly suited for safe storage in hazardous environments. Featuring thermostat and compressor components that are isolated within a vapor-proof enclosure, and electrical connections that are hardwired, these units will prevent sparking and accidental disconnection. All models feature manual defrost.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Min Temperature - Refrigerator (° F) Power (VAC) Refrigerator Capacity (cu ft)
Availability Pricing
Thermo Scientific 05EREETSA Explosion Proof Undercounter Refrigerator, 5.5 cu ft; 115V EW-44200-20 Mfr # 05EREETSA
COMPARE
33 115 5.5
Call For Price
Thermo Scientific 3566A Explosion-Proof Refrigerator, 20ft3 with 1 door EW-44200-25 Mfr # 3566A
COMPARE
115
Call For Price
Thermo Scientific 3557-4 Undercounter explosion-proof refrigerator/freezer, 5.6ft3 EW-44200-95 Mfr # 3557-4
COMPARE
115
Call For Price
Thermo Scientific 3556 Undercounter Refrigerators; Description, Flammable material storage; 120V EW-44201-20 Mfr # 3556
COMPARE
115
Call For Price
Thermo Scientific 3566-10A Flammable Material Storage Refrigerator, 20 cu ft; 115V/60Hz EW-44201-30 Mfr # 3566-10A
COMPARE
220
Call For Price
Thermo Scientific Explosion-Proof Refrigerator, 20 cu ft; 115 V EW-44203-25 Mfr # 20EREETSA
COMPARE
33 115 20
Call For Price
Thermo Scientific Flammable-Material Upright Refrigerator, 20 cu ft; 115 V EW-44204-30 Mfr # 20FREETSA
COMPARE
33 115 20
Call For Price
Showing 1- 7 of 7