Thermo Scientific Heraeus Megafuge 16 and 16R Centrifuges
Designed around you and your applications
  • AutoLock® III rotor system provides easy rotor change in about 3-seconds!
  • Motorized lid-latch reduces wear and closure time
  • Intuitive display and control for routine and repetitive environments — perfect for clinical protocols, cell culture applications, and microplate processing
These general-purpose centrifuges provide flexibility with an easy change rotor system, and improved safety features. Unique, AutoLock® rotor system makes changes of rotors and buckets easy. With the push of a button, change the rotor — no need for tools. SMARTSpin® rotor management system maximizes safety and separation by optimizing acceleration, braking, and residual load imbalance for any rotor and bucket option.
The compact footprint minimizes the lab space required, while the sleek design provides improved ergonomics and easy clean-up. Motorized lid latch requires minimal effort for closing. Six program keys allow for easier recall of frequently used settings to save time.
$7,422.00 - $11,090.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Refrigerated
Availability Pricing
Thermo Scientific Heraeus 75004231 Centrifuge; 120 VAC, 60 Hz EW-17707-41 Mfr # 75004231
COMPARE
120 No
$7,422.00USD / Each
Thermo Scientific Heraeus 75004230 Centrifuge; 230 VAC, 50/60 Hz EW-17707-42 Mfr # 75004230
COMPARE
230 No
$7,504.00USD / Each
Thermo Scientific Heraeus Megafuge 16R Refrigerated Centrifuge; 120 VAC EW-17707-51 Mfr # 75004271
COMPARE
120 Yes
$11,090.00USD / Each
Thermo Scientific Heraeus Megafuge 16R Refrigerated Centrifuge; 230 VAC EW-17707-52 Mfr # 75004270
COMPARE
230 Yes
$11,090.00USD / Each