Thermo Scientific Heraeus Megafuge 40/40R Centrifuges
4-liter capacity in a 3-liter footprint delivers high throughput and exceptional performance
  • AutoLock® III rotor system provides easy rotor change in about 3-seconds!
  • Motorized lid-latch reduces wear and closure time
  • High sample capacity and higher throughput for up to 40% in energy savings
  • Adjustable acceleration and deceleration rates, as well as pre-selection of speed or RCF
These general-purpose centrifuges provide industry-leading capacity, flexibility with an easy change rotor system, and improved safety features. Unique, AutoLock® rotor system makes changes of rotors and buckets easy. With the push of a button, change the rotor and process parameters — no need for tools. SMARTSpin® rotor management system maximizes safety and separation by optimizing acceleration, braking, and residual load imbalance for any rotor and bucket option.
Compact footprint minimizes the lab space required, while the sleek design provides improved ergonomics and easy clean-up. Motorized lid latch requires minimal effort for closing. Six program keys allow for easier recall of frequently used settings to save time.
$10,282.00 - $15,632.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Refrigerated
Availability Pricing
Thermo Scientific Heraeus 75004504 Centrifuge, 4L, 120VAC EW-17707-20 Mfr # 75004504
COMPARE
120 No
$10,282.00USD / Each
Thermo Scientific Heraeus Megafuge 40 Centrifuge, 4L, 230VAC EW-17707-25 Mfr # 75004503
COMPARE
230 No
$10,282.00USD / Each
Thermo Scientific Heraeus Megafuge 40R Refrigerated Centrifuge, 120 VAC EW-17707-30 Mfr # 75004519
COMPARE
120 Yes
$15,632.00USD / Each
Thermo Scientific Heraeus Megafuge 40R Refrigerated Centrifuge, 230 VAC EW-17707-35 Mfr # 75004518
COMPARE
230 Yes
$15,632.00USD / Each