Thermo Scientific Heraeus Megafuge 8 Centrifuge Rotors
Exceptional capacity and flexible processing adds value to any lab
  • SMARTSpin imbalance detection system prevents hazardous sample running
  • Quality design and brushless drive ensure years of quiet use
  • Biocontainment option keeps you safe
Conforming to the latest safety standards, the Megafuge 8 is a compact bench centrifuge that satisfies many sample running options. The secure push-button Auto-Lock® rotor exchange system with glove-friendly design allows you to change out rotors with ease, requiring only one hand! The SMARTSpin imbalance detection system optimizes acceleration, braking, and load imbalance for smooth runs and reproducible separations.
Centrifuge features one-touch operation and pre-saved programming with password protection. The control panel with highly visible backlit display is glove- and detergent-friendly. Brushless motor provides quiet performance and a stress free work environment.
Rotors require adapters, order separately.
$1,856.00 - $2,868.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Tube Capacity
Availability Pricing
Thermo Scientific Heraeus 75005765 MicroClick Centrifuge Rotor, 18 x 5 mL EW-17701-29 Mfr # 75005765
COMPARE
5500 18 x 5 mL
$1,856.00USD / Each
Thermo Scientific Heraeus TX-150 Swinging Bucket Rotor, 4 x 145 mL, unsealed, requires Adapter EW-17701-30 Mfr # 75005701
COMPARE
4500 4 x 145 mL
$2,232.00USD / Each
Thermo Scientific Heraeus 75005706 M10 Microplate Swinging Bucket Rotor, Rma x 139mm, requires Adapter EW-17701-40 Mfr # 75005706
COMPARE
4400 Bucket is sold separately-4 x standard microplates or -2 x deepwell microplates
$2,868.00USD / Each