Thermo Scientific Heraeus Megafuge 8 Centrifuge Tube Adapters for Microplate Swinging Bucket Rotor
Exceptional capacity and flexible processing adds value to any lab
  • SMARTSpin imbalance detection system prevents hazardous sample running
  • Quality design and brushless drive ensure years of quiet use
  • Biocontainment option keeps you safe
Conforming to the latest safety standards, the Megafuge 8 is a compact bench centrifuge that satisfies many sample running options. The secure push-button Auto-Lock® rotor exchange system with glove-friendly design allows you to change out rotors with ease, requiring only one hand! The SMARTSpin imbalance detection system optimizes acceleration, braking, and load imbalance for smooth runs and reproducible separations.
Centrifuge features one-touch operation and pre-saved programming with password protection. The control panel with highly visible backlit display is glove- and detergent-friendly. Brushless motor provides quiet performance and a stress free work environment.
$1,142.00 - $1,606.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific Heraeus M10 Centrifuge Tube Adapter, Unsealed Bucket for; 2/Pk EW-17701-41 Mfr # 75005723
COMPARE
CentrifugeTubeAdapter,UnsealedBucketfor;2/Pk
$1,142.00USD / Each
Thermo Scientific Heraeus 75005721 Centrifuge Tube Adapter, Sealed Bucket for M10; 2/Pk EW-17701-42 Mfr # 75005721
COMPARE
CentrifugeTubeAdapter,SealedBucketforM10;2/Pk
$1,606.00USD / Each