Thermo Scientific Heraeus Pico and Fresco Microcentrifuges
Powerful centrifugation in a small footprint
  • Generous sample capacity in a quiet compact unit
These high-speed, compact microcentrifuges provide efficient sample processing and a generous sample capacity. Quiet motor with fast acceleration and deceleration allows you to minimize noise and processing time. Units feature a sleek, lightweight design for easy clean-up and minimal benchtop utilization. LED controls provide easy set-up and operation.
$2,884.00 - $9,120.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Refrigerated Tube Capacity
Availability Pricing
Thermo Scientific Heraeus Pico 17 MicroCentrifuge, 24-Pl Rotor; 120V EW-17703-00 Mfr # 75002411
COMPARE
120 No 24 x 1.5/2.0 mL-Other rotors are available
$3,144.00USD / Each
Thermo Scientific Heraeus Pico 17 MicroCentrifuge, 24-Pl Rotor; 230 VAC EW-17703-05 Mfr # 75002410
COMPARE
230 No 24 x 1.5/2.0 mL-Other rotors are available
$2,884.00USD / Each
Thermo Scientific Heraeus Fresco 17 Refrigerated Micro Centrifuge, 24-Pl Rotor; 120V EW-17703-10 Mfr # 75002421
COMPARE
120 Yes 24 x 1.5/2.0 mL-Other rotors are available
$8,126.00USD / Each
Thermo Scientific Heraeus Fresco 17 Refrigerated Micro Centrifuge, 24-Pl Rotor; 230 VAC EW-17703-15 Mfr # 75002420
COMPARE
230 Yes 24 x 1.5/2.0 mL-Other rotors are available
$8,126.00USD / Each
Thermo Scientific Heraeus Pico 21 MicroCentrifuge, 24-Pl Rotor; 120V EW-17703-20 Mfr # 75002416
COMPARE
120 No 24 x 1.5/2.0 mL-Other rotors are available
$3,944.00USD / Each
Thermo Scientific Heraeus Pico 21 MicroCentrifuge, 24-Pl Rotor; 230 VAC EW-17703-25 Mfr # 75002415
COMPARE
230 No 24 x 1.5/2.0 mL-Other rotors are available
$3,414.00USD / Each
Thermo Scientific Heraeus Fresco 21 Refrigerated Micro Centrifuge, 24-Pl Rotor; 120V EW-17703-30 Mfr # 75002426
COMPARE
120 Yes 24 x 1.5/2.0 mL-Other rotors are available
$9,120.00USD / Each
Thermo Scientific Heraeus Fresco 21 Refrigerated Micro Centrifuge, 24-Pl Rotor; 230 VAC EW-17703-35 Mfr # 75002425
COMPARE
230 Yes 24 x 1.5/2.0 mL-Other rotors are available
$9,120.00USD / Each
Thermo Scientific Heraeus Pico 17 MicroCentrifuge w/Hematocrit Rotor; 115 VAC EW-17703-40 Mfr # 75002492
COMPARE
120 No 24 x Hematocrit tubes-Other rotors are available
$3,214.00USD / Each
Thermo Scientific Heraeus Pico 17 MicroCentrifuge w/Hematocrit Rotor; 230 VAC EW-17703-45 Mfr # 75002491
COMPARE
230 No 24 x Hematocrit tubes-Other rotors are available
$3,252.00USD / Each
Thermo Scientific Heraeus Pico 21 MicroCentrifuge w/2 Rotors; 115 VAC EW-17703-50 Mfr # 75002477
COMPARE
120 No 24 x 1.5/2.0 mL-18 x 0.5/2.0 mL dual row rotor-Other rotors are available
$3,994.00USD / Each
Thermo Scientific Heraeus Pico 21 MicroCentrifuge w/2 Rotors; 230 VAC EW-17703-55 Mfr # 75002475
COMPARE
230 No 24 x 1.5/2.0 mL-18 x 0.5/2.0 mL dual row rotor-Other rotors are available
$4,144.00USD / Each
Thermo Scientific Heraeus Fresco 21 MicroCentrifuge w/2 Rotors; 115 VAC EW-17703-60 Mfr # 75002478
COMPARE
120 Yes 24 x 1.5/2.0 mL-18 x 0.5/2.0 mL dual row rotor-Other rotors are available
$8,130.00USD / Each
Thermo Scientific Heraeus Fresco 21 MicroCentrifuge w/2 Rotors; 230 VAC EW-17703-65 Mfr # 75002476
COMPARE
230 Yes 24 x 1.5/2.0 mL-18 x 0.5/2.0 mL dual row rotor-Other rotors are available
$8,956.00USD / Each
Showing 1- 14 of 14