Thermo Scientific Heratherm Advanced Protocol Security (APS) Gravity Convection Ovens
Extra layer of protection when process reliability and security are most important
  • Automatic undertemperature alarm ensures samples are kept at the correct temperature
  • Door lock prevents disruption, tampering, or accidental opening
  • Door alarm notifies when door is accidentally left open
  • Models with robust stainless steel exterior are ideal for pharmaceutical labs
Security gravity convection ovens offer additional safety features for essential sample protection. A gentle air circulation pattern with low turbulence ensures accurate, even heating from 50 to 330°C. Programmable controller with large easy-to-view display and touch-button controls can store up to 10 programs with 10 discrete steps. Timer function can be set to run on weekly, hourly, or real time.
Auto-dry function deactivates the oven when samples are dry, saving energy. Additional features include a socket for an independent sensor that when connected is shown on display, an electronically controlled damper, a boost feature for rapid heating of cold oven, and a dedicated access port for the introduction of external temperature sensors.
Smooth stainless steel inner chamber has rounded corners for easy cleaning. The door opens 180°, providing easy access to interior for cleaning and loading of probes. A standard overtemperature alarm and additional undertemperature alarm ensures your samples are always protected. The RS-232 interface lets you connect to your computer for documentation of parameters.
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Thermo Scientific Heratherm 51028118 APS Gravity Oven; 2 cu ft, coated steel, 120V EW-38800-54 Mfr # 51028118
COMPARE
2.15 120
$3,363.50USD / Each
Thermo Scientific Heratherm 51028120 APS Gravity Oven; 6.1 cu ft, coated steel, 208-240V EW-38800-58 Mfr # 51028120
COMPARE
6.05 208 / 240
$4,370.50USD / Each
Thermo Scientific Heratherm 51028537 APS Gravity Oven; 3.5 cu ft, SS exterior, 208-240V EW-38800-62 Mfr # 51028537
COMPARE
3.6 208 / 240
$3,966.50USD / Each
Thermo Scientific Heratherm 51028538 APS Gravity Oven; 6.1 cu ft, SS exterior, 208-240V EW-38800-64 Mfr # 51028538
COMPARE
6.05 208 / 240
$4,702.50USD / Each