This product is now discontinued, View Related Products
Thermo Scientific Heratherm Advanced Protocol Security Incubators
Offers unique 140°C decontamination routine, providing safety for samples!
  • Dual convection for application versatility — adjustable fan speed from 10 to 100%
  • Advanced digital timer for daily or weekly on/off cycles
  • Additional safety features for ultimate sample protection
Incubators include additional safety features for essential sample protection: a lockable door for restricted access, an alarm sounds if the door is left open accidentally, and an automatic over-and under-temperature alarm. Broad temperature range from 5°C above ambient to 105°C is suitable for drying applications. One-of-a-kind 140°C contamination cycle reduces microorganism to a minimum — comparable to sterilization — within a six-hour heating cycle. With the 140°C contamination cycle there is no need for separate autoclaving of internal fittings. Units have an easy-to-clean stainless steel interior, large easy-to-read vacuum fluorescent display, and internal glass door to allow sample viewing without impacting temperature.
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters) Max Temperature (° C) Power (VAC)
Availability Pricing
Thermo Scientific Heratherm IMH60-S Advanced Security Incubator, 2 cu ft, Dual; 120V EW-38800-14 Mfr # 51028069
COMPARE
66 105 120
Call For Price
Thermo Scientific Heratherm IMH400-S Advanced Security Incubator, 13.5 cu ft, Dual; 120V EW-38800-15 Mfr # 51029323
COMPARE
382 105 120
Call For Price
Thermo Scientific Heratherm IMH100-S Advanced Security Incubator, 3.5 cu ft, Dual; 120V EW-38800-16 Mfr # 51028070
COMPARE
104 105 120
Call For Price
Thermo Scientific Heratherm IMH750-S Advanced Security Incubator, 24.8 cu ft, Dual; 120V EW-38800-17 Mfr # 51029335
COMPARE
702 105 120
Call For Price
Thermo Scientific Heratherm IMH180-S Advanced Security Incubator, 6.4 cu ft, Dual; 120V EW-38800-18 Mfr # 51028111
COMPARE
178 105 120
Call For Price
Thermo Scientific Heratherm 51028264 Advanced Security Incubator, 2 cu ft SS, Dual; 120V EW-38800-20 Mfr # 51028264
COMPARE
66 105 120
Call For Price
Thermo Scientific Heratherm 51029324 Advanced Security Incubator, 13.5 cu ft SS, Dual; 120V EW-38800-21 Mfr # 51029324
COMPARE
382 105 120
Call For Price
Thermo Scientific Heratherm 51028535 Advanced Security Incubator, 3.5 cu ft SS, Dual; 120V EW-38800-22 Mfr # 51028535
COMPARE
104 105 120
Call For Price
Thermo Scientific Heratherm 51029336 Advanced Security Incubator, 24.8 cu ft SS, Dual; 120V EW-38800-23 Mfr # 51029336
COMPARE
702 105 120
Call For Price
Thermo Scientific Heratherm 51028327 Advanced Security Incubator, 6.4 cu ft SS, Dual; 120V EW-38800-24 Mfr # 51028327
COMPARE
178 105 120
Call For Price
Showing 1- 10 of 10