Thermo Scientific Hybridization Bottle and Mesh System
Manufactured using thick walled borosilicate glass to ensure safe and easy handling
  • Improves the flow of probe solution around and through membranes for optimal results
  • Designed to exacting safety standards, these bottles have screw caps containing flat PTFE seals to ensure leakresistant hybridization
  • Full range of bottles accommodates every size of membrane
  • Enables multiple blots to be processed simultaneously in one 35mm diameter bottle without direct overlap
Choose from bottles, mesh, and bottle grippers to complete your hybridization and mesh system. Bottle grippers are ideal for safe and easy handling of bottles.
$235.00USD / Pkg of 5
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific 222053 Oven Mesh Sheets: Large, 23 x 23cm 5/PK EW-13247-32 Mfr # 222053
COMPARE
Oven Mesh Sheets: Large, 23 x 23cm 5/PK
$235.00USD / Pkg of 5