This product is now discontinued, View Related Products
Thermo Scientific Hypersil GOLD PFP (perfluorophenyl) Columns
Thermo Scientific Hypersil GOLD PFP Columns provide excellent peak shape and selectivity of fluorinated and other halogenated compounds. These columns are also particularly useful for non-halogenated aromatic compounds containing hydroxyl, carboxyl, nitro, or other polar groups; or in separating difficult mixtures containing closely related halogenated and polar compounds. Stable in a pH range of 2 to 8.
Call For Price
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil 25402-021030 GOLD PFP Column - Length 20mm, 1.0mm ID, 1.9µm particle size EW-01882-04 Mfr # 25402-021030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-022130 GOLD PFP Column - Length 20mm, 2.1mm ID, 1.9µm particle size EW-01882-05 Mfr # 25402-022130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-023030 GOLD PFP Column - Length 20mm, 3.0mm ID, 1.9µm particle size EW-01882-06 Mfr # 25402-023030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-031030 GOLD PFP Column - Length 30mm, 1.0mm ID, 1.9µm particle size EW-01882-07 Mfr # 25402-031030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-032130 GOLD PFP Column - Length 30mm, 2.1mm ID, 1.9µm particle size EW-01882-08 Mfr # 25402-032130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-033030 GOLD PFP Column - Length 30mm, 3.0mm ID, 1.9µm particle size EW-01882-09 Mfr # 25402-033030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-051030 GOLD PFP Column - Length 50mm, 1.0mm ID, 1.9µm particle size EW-01882-11 Mfr # 25402-051030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-052130 GOLD PFP Column - Length 50mm, 2.1mm ID, 1.9µm particle size EW-01882-14 Mfr # 25402-052130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-053030 GOLD PFP Column - Length 50mm, 3.0mm ID, 1.9µm particle size EW-01882-15 Mfr # 25402-053030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-101030 GOLD PFP Column - Length 100mm, 1.0mm ID, 1.9µm particle size EW-01882-21 Mfr # 25402-101030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-102130 GOLD PFP Column - Length 100mm, 2.1mm ID, 1.9µm particle size EW-01882-22 Mfr # 25402-102130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-103030 GOLD PFP Column - Length 100mm, 3.0mm ID, 1.9µm particle size EW-01882-23 Mfr # 25402-103030
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-152130 GOLD PFP Column - Length 150mm, 3.0mm ID, 1.9µm particle size EW-01882-24 Mfr # 25402-152130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25402-202130 GOLD PFP Column - Length 200mm, 3.0mm ID, 1.9µm particle size EW-01882-25 Mfr # 25402-202130
COMPARE
1.9
Call For Price
Thermo Scientific Hypersil 25403-031030 GOLD PFP Column - Length 30mm, 1.0mm ID, 3µm particle size EW-01882-33 Mfr # 25403-031030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25403-032130 GOLD PFP Column - Length 30mm, 2.1mm ID, 3µm particle size EW-01882-34 Mfr # 25403-032130
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25403-033030 GOLD PFP Column - Length 30mm, 3.0mm ID, 3µm particle size EW-01882-35 Mfr # 25403-033030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25403-034030 GOLD PFP Column - Length 30mm, 4.0mm ID, 3µm particle size EW-01882-36 Mfr # 25403-034030
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25403-034630 GOLD PFP Column - Length 30mm, 4.6mm ID, 3µm particle size EW-01882-37 Mfr # 25403-034630
COMPARE
3
Call For Price
Thermo Scientific Hypersil 25403-051030 GOLD PFP Column - Length 50mm, 1.0mm ID, 3µm particle size EW-01882-41 Mfr # 25403-051030
COMPARE
3
Call For Price
Showing 1 - 20 of 50