Thermo Scientific™ Hypersil™ SAS Classical C1 HPLC Columns
High-efficiency columns with proven results and reproducibility
  • Suitable for SAS (C1) phase
Thermo Scientific™ Hypersil™ columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil SAS (C1) Columns are the least retentive of the Hypersil columns and are useful for ion-pair separations. Exhibiting good stability, these columns show selectivity for polar analytes and multifunctional compounds. Stable in pH range of 2 to 8.
$657.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil SAS HPLC Column, C1, 3 µm, 250 mm L x 4.6 mm ID EW-01888-56 Mfr # 30503-254630
COMPARE
4.6 250 3
$657.00USD / Each