Thermo Scientific Lindberg Blue M Three-Zone Tube Furnaces
Temperature uniformity ±1.0°C over 24 inches!
  • Control each zone independently
  • Accept multiple tube sizes for versatility
These three-zone tube furnaces feature three 16-segment programmable controllers—one for each zone. Microprocessor-based self-tuning PID control provides excellent control without over-shooting of set temperature. Protect your samples with the adjustable high limit overtemperature protection feature. Furnaces have a uniform temperature throughout the length of the process tube and excellent radial and linear temperature uniformity. The digital LED displays your set and actual temperatures at the same time.
Order process tubes separately.
$10,200.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Process Tube OD (in)
Availability Pricing
Thermo Scientific Lindberg STF55346C Blue M Three-Zone Tube Furnace, 1"-3" OD; 208/240 VAC EW-33851-25 Mfr # STF55346C-1
COMPARE
208 / 240 1 to 3
$10,200.00USD / Each