Thermo Scientific Nalgene 180 Clear PVC Tubing
Thermo Scientific Nalgene 180 clear PVC tubing is ideal for applications in pharmaceutical, biotechnology, and general laboratory use. Resistant to hardening and discoloration, it offers good chemical resistance. The PVC resin meets USP Class VI and USDA requirements and may be sterilized either by autoclaving or with ethylene oxide. Usable from -26 to 160deg F (-32 to 71deg C).
$38.50 - $672.00USD / Pkg of 1
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
Thermo Scientific Nalgene 8000-0010 180 PVC Tubing, 1/8 X 3/16", 50' Per Pack
Buy More Save More
EW-96700-04 Mfr # 8000-0010
COMPARE
34 1/8 3/16
$38.50USD / Pkg of 1
Thermo Scientific Nalgene 8000-0020 180 PVC Tubing, 1/8 X 1/4", 50' Per Pack
Buy More Save More
EW-96700-05 Mfr # 8000-0020
COMPARE
50 1/8 1/4
$83.50USD / Pkg of 1
Thermo Scientific Nalgene 8000-0030 180 PVC Tubing, 3/16 X 5/16", 50' Per Pack
Buy More Save More
EW-96700-07 Mfr # 8000-0030
COMPARE
40 3/16 5/16
$114.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0060 180 PVC Tubing, 1/4 X 3/8", 50' Per Pack
Buy More Save More
EW-96700-08 Mfr # 8000-0060
COMPARE
34 1/4 3/8
$144.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0070 180 PVC Tubing, 1/4 X 7/16", 50' Per Pack
Buy More Save More
EW-96700-09 Mfr # 8000-0070
COMPARE
44 1/4 7/16
$231.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0080 180 PVC Tubing, 1/4 X 1/2", 50' Per Pack
Buy More Save More
EW-96700-10 Mfr # 8000-0080
COMPARE
52 1/4 1/2
$271.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0090 180 PVC Tubing, 5/16 X 7/16", 50' Per Pack
Buy More Save More
EW-96700-11 Mfr # 8000-0090
COMPARE
30 5/16 9/16
$161.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0110 180 PVC Tubing, 5/16 X 9/16", 50' Per Pack
Buy More Save More
EW-96700-13 Mfr # 8000-0110
COMPARE
46 5/16 9/16
$285.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0120 180 PVC Tubing, 3/8 X 1/2", 50' Per Pack
Buy More Save More
EW-96700-14 Mfr # 8000-0120
COMPARE
26 3/8 1/2
$186.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0130 180 PVC Tubing, 3/8 X 9/16", 50' Per Pack
Buy More Save More
EW-96700-15 Mfr # 8000-0130
COMPARE
36 3/8 9/16
$243.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0180 180 PVC Tubing, 1/2 X 5/8", 50' Per Pack
Buy More Save More
EW-96700-18 Mfr # 8000-0180
COMPARE
22 1/2 5/8
$229.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0200 180 PVC Tubing, 1/2 X 3/4", 50' Per Pack
Buy More Save More
EW-96700-20 Mfr # 8000-0200
COMPARE
34 1/2 3/4
$421.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0260 180 PVC Tubing, 5/8 X 7/8", 50' Per Pack
Buy More Save More
EW-96700-21 Mfr # 8000-0260
COMPARE
28 5/8 7/8
$478.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0310 180 PVC Tubing, 3/4 X 1", 50' Per Pack
Buy More Save More
EW-96700-22 Mfr # 8000-0310
COMPARE
26 3/4 1
$544.00USD / Pkg of 1
Thermo Scientific Nalgene 8000-0350 180 PVC Tubing, 1 X 1-1/4", 50' Per Pack
Buy More Save More
EW-96700-23 Mfr # 8000-0350
COMPARE
22 1 1 1/4
$672.00USD / Pkg of 1
Showing 1- 15 of 15