Thermo Scientific Nalgene Polypropylene Copolymer (PPCO) Dispensing Bottles
Easily dispenses one drop at a time
  • Easy viewing of liquid samples
  • Autoclavable
Use these leakproof PPCO bottles to dispense sterile solutions; squeeze for fine, steady stream. Bottles can be autoclaved full, with tips uncapped. The tethered spout cap snaps securely into place and won't pop off or leak when the closed bottle is squeezed.
$116.00 - $130.00USD / Pkg of 6
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap Material Material
Availability Pricing
Thermo Scientific Nalgene Polypropylene Copolymer Dispensing Bottle, 125 mL EW-06086-45 Mfr # DS2420-0125
COMPARE
125 Polypropylene (PP) Polypropylene Copolyment (PPCO)
$116.00USD / Pkg of 6
Thermo Scientific Nalgene Polypropylene Copolymer Dispensing Bottle, 250 mL EW-06086-55 Mfr # DS2420-0250
COMPARE
250 Polypropylene (PP) Polypropylene Copolyment (PPCO)
$130.00USD / Pkg of 6