Thermo Scientific Nalgene Conical Centrifuge Bottles, 175 mL
Get better separation of large samples
  • Measure more precisely with molded-in graduations
  • Leakproof
The bottles are available in PPCO or PC. Feature molded-in graduations and PP sealing cap. Must use adapter 06106-78 to fit most rotors.
$176.00USD / Pkg of 4
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Tube Bottom Shape Tube Capacity (mL) Tube Material
Availability Pricing
Thermo Scientific Nalgene 3143-0175 Centrifuge bottle; 175 mL; PPCO; 4/pack
Buy More Save More
EW-06107-28 Mfr # 3143-0175
COMPARE
Conical 175 Polypropylene
$176.00USD / Pkg of 4