Thermo Scientific Nalgene Filling/Venting Caps
Transfer aseptic liquid safely
  • Autoclavable
Filling/venting caps have thermoplastic elastomer (TPE) gaskets to allow aseptic liquid transfer.
Include 6" (15 cm) and 30" (76 cm) lengths of platinum-cured silicone tubing.
$85.00 - $95.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Material Tubing ID
Availability Pricing
Thermo Scientific Nalgene Filling/Venting Cap for 1/2" Tubing, 83B
Buy More Save More
EW-06258-00 Mfr # 2162-0830
COMPARE
83B Polypropylene 12.7 mm
$95.50USD / Each
Thermo Scientific Nalgene Filling/Venting Cap for 1/4" Tubing, 53B
Buy More Save More
EW-06258-10 Mfr # 2162-0531
COMPARE
53B Polypropylene 6.4 mm
$85.00USD / Each
Thermo Scientific Nalgene Filling/Venting Cap for 1/4" Tubing, 83B
Buy More Save More
EW-06258-20 Mfr # 2162-0831
COMPARE
83B Polypropylene 6.4 mm
$95.00USD / Each