Thermo Scientific Nalgene High-Density Polyethylene (HDPE) Wide-Mouth Bottles
Useful for storing dry chemicals or for taking water samples. May be used for freezer storage down to -148°>F (-100°>C). Bottles include polypropylene caps; neck diameter is 89 mm. Order replacement caps separately.
$29.00 - $38.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Material Overall Height (in)
Availability Pricing
Thermo Scientific Nalgene 2120-0005 Wide-Mouth Bottle, HDPE, 2 L
Buy More Save More
EW-06055-10 Mfr # 2120-0005
COMPARE
2 HDPE 9
$29.00USD / Each
Thermo Scientific Nalgene 2120-0010 Wide-Mouth Bottle, HDPE, 4 L
Buy More Save More
EW-06055-20 Mfr # 2120-0010
COMPARE
4 HDPE 11
$38.00USD / Each