This product is now discontinued, View Related Products
Thermo Scientific Nalgene Low-Density Polypropylene Dropping Bottles with Removable Pipettes
Dropping pipettes may be removed without unscrewing cap. Assembled unit includes narrow-mouth bottle and dropping pipette with polypropylene (PP) screw closure. The 30-mL size has a dropping pipette made completely of polyethylene (PE). Other sizes have polyethylene dropper with neoprene bulb. Nalgene(R).
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Thermo Scientific Nalgene 2416-0030 low-density polyethylene dropping bottle with pipette, 30 mL EW-06090-10 Mfr # 2416-0030
COMPARE
30 Low Density Polyethyene (LDPE)
Call For Price
Thermo Scientific Nalgene 2416-0060 LDPE Dropping Bottle w/ Pippette, 60 mL EW-06090-20 Mfr # 2416-0060
COMPARE
60 Low Density Polyethyene (LDPE)
Call For Price
Thermo Scientific Nalgene 2416-0125 LDPE Dropping Bottle w/ Pippette, 125 mL EW-06090-40 Mfr # 2416-0125
COMPARE
125 Low Density Polyethyene (LDPE)