Thermo Scientific™ Nalgene™ PFA Graduated Beakers with Handle
Sturdy easy-grip handles
  • Rigid and translucent, enabling easy and consistent viewing of liquid levels
  • Molded-in graduation in both oz. and mL won't wear off with continuous use or washings
  • Built-in rectangular handle for easy, safe handling and pouring
  • Autoclavable
Conduct high-purity applications that use strong oxidizing agents, acids, hydrocarbons and ketones with Thermo Scientific™ Nalgene™ PFA Graduated Beakers with Handles. These beakers feature extraordinary chemical resistance, making them easy to clean and resistant to contamination. Beakers can withstand exposure to extreme temperatures from -270° to +250°C.
$665.00 - $808.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Thermo Scientific Nalgene DS1511-1000 Beaker w/handle, PFA, 1000 mL, 1/ea EW-06006-00 Mfr # DS1511-1000
COMPARE
1000 50 Perfluoroalkoxy (PFA)
$665.00USD / Each
Thermo Scientific Nalgene DS1511-3000 Beaker w/handle, PFA, 3000 mL, 1/ea EW-06006-05 Mfr # DS1511-3000
COMPARE
3000 125 Perfluoroalkoxy (PFA)
$808.00USD / Each