Thermo Scientific Nalgene Polyethersulfone (PES) Filter Units and Bottletop Filters
  • Hydrophilic PES provides fast flow rate
  • Low protein binding — excellent for filtering solutions for cell culture applications
Thermo Scientific Nalgene® Bottletop Filters are polystyrene funnels with integrally welded filter membranes designed for direct filtration into sterile glass media bottles. They thread securely onto bottles with 45-mm neck. Side arms with quick-disconnect tubing adapters simplify vacuum line attachment.
$75.00 - $755.00USD / Pkg of 12
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Pore Size (µm) Sterile
Availability Pricing
Thermo Scientific Nalgene 524-0020 Filter Units, Gridded 0.2um PES, 115 mL; 72/Pk EW-06730-25 Mfr # 524-0020
COMPARE
115 0.2 Yes
$755.00USD / Pkg of 12
Thermo Scientific Nalgene 565-0020 Filter Units, 0.2um PES, 150 mL; 12/Pk EW-06730-31 Mfr # 565-0020
COMPARE
150 0.2 Yes
$162.00USD / Pkg of 12
Thermo Scientific Nalgene 165-0045 Filter Units, 0.45um PES, 150 mL; 12/Pk EW-06730-32 Mfr # 165-0045
COMPARE
150 0.45 Yes
$153.00USD / Pkg of 12
Thermo Scientific Nalgene 124-0045 Filter Units, 0.45um PES, 115 mL; 12/Pk EW-06730-34 Mfr # 124-0045
COMPARE
115 0.45 Yes
$172.00USD / Pkg of 12
Thermo Scientific Nalgene 566-0020 Filter Units, 0.2um PES, 500 mL; 12/Pk EW-06730-43 Mfr # 566-0020
COMPARE
500 0.2 Yes
$335.00USD / Pkg of 12
Thermo Scientific Nalgene 166-0045 Filter Units, 0.45um PES, 500 mL; 12/Pk EW-06730-54 Mfr # 166-0045
COMPARE
500 0.45 Yes
$314.00USD / Pkg of 12
Thermo Scientific Nalgene 567-0020 Filter Units, 0.20um PES, 1000 mL; 12/Pk EW-06730-56 Mfr # 567-0020
COMPARE
1000 0.2 Yes
$433.00USD / Pkg of 12
Thermo Scientific Nalgene 167-0045 Filter Units, 0.45um PES, 1000 mL; 12/Pk EW-06730-58 Mfr # 167-0045
COMPARE
1000 0.45 Yes
$404.00USD / Pkg of 12
Thermo Scientific Nalgene 596-4520 Bottletop Filters, 0.2um PES, 150 mL; 12/Pk EW-06731-11 Mfr # 596-4520
COMPARE
150 0.2 Yes
$153.00USD / Pkg of 12
Thermo Scientific Nalgene 296-4545 Polyethersulfone (PES) Bottletop Filters; 150 mL, 0.45 µm Pore Size EW-06731-12 Mfr # 296-4545
COMPARE
150 0.45 Yes
$75.00USD / Pkg
Thermo Scientific Nalgene 595-4520 Bottletop Filters, 0.2um PES, 500 mL; 12/Pk EW-06731-15 Mfr # 595-4520
COMPARE
500 0.2 Yes
$225.00USD / Pkg of 12
Thermo Scientific Nalgene 295-4545 Bottletop Filters, 0.45um PES, 500 mL; 12/Pk EW-06731-16 Mfr # 295-4545
COMPARE
500 0.45 Yes
$208.00USD / Pkg of 12
Thermo Scientific Nalgene 597-4520 Bottletop Filters, 0.2um PES, 1000 mL; 12/Pk EW-06731-18 Mfr # 597-4520
COMPARE
1000 0.2 Yes
$323.00USD / Pkg of 12
Thermo Scientific FILTER NYL 250ML 0.45UM 12/CS EW-06731-86 Mfr # 153-0045
COMPARE
$264.00USD / Case
Thermo Scientific FILTER NYL 500ML 0.45UM 12/CS EW-06731-90 Mfr # 151-4045
COMPARE
$373.00USD / Case
Showing 1- 15 of 15
Need Help with this product?
Call Us At1-847-549-7600
Live chat