Thermo Scientific Nalgene Polypropylene Wide-Mouth Bottles
Useful for storing dry chemicals or for taking water samples. May be used for freezer storage down to -148°>F (-100°>C). Bottles include polypropylene caps; neck diameter is 89 mm. Order replacement caps separately.
$32.00 - $41.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Material Overall Height (in)
Availability Pricing
COMPARE
2 Polypropylene 9
$32.00USD / Each
COMPARE
4 Polypropylene 11
$41.00USD / Each