Thermo Scientific Nalgene Surfactant-Free Cellulose Acetate (SFCA) Sterile Filter Units and Bottletop Filters
Contain no wetting agents, which can affect sensitive cells
  • Exhibit low protein binding and excellent flow rates
$159.00 - $452.00USD / Pkg of 12
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Pore Size (µm) Sterile
Availability Pricing
Thermo Scientific Nalgene 155-0020 Filter Units, 0.2um SFCA, 150 mL; 12/Pk EW-06731-20 Mfr # 155-0020
COMPARE
150 0.2 Yes
$197.00USD / Pkg of 12
Thermo Scientific Nalgene 155-0045 Filter Units, 0.45um SFCA, 150 mL; 12/Pk EW-06731-22 Mfr # 155-0045
COMPARE
150 0.45 Yes
$197.00USD / Pkg of 12
Thermo Scientific Nalgene 156-4020 Filter Units, 0.2um SFCA, 500 mL; 12/Pk EW-06731-24 Mfr # 156-4020
COMPARE
500 0.2 Yes
$309.00USD / Pkg of 12
Thermo Scientific Nalgene 156-4045 Filter Units, 0.45um SFCA, 500 mL; 12/Pk EW-06731-26 Mfr # 156-4045
COMPARE
500 0.45 Yes
$309.00USD / Pkg of 12
Thermo Scientific Nalgene 161-0020 Filter Units, 0.2um SFCA, 1000 mL; 12/Pk EW-06731-27 Mfr # 161-0020
COMPARE
1000 0.2 Yes
$452.00USD / Pkg of 12
Thermo Scientific Nalgene 161-0045 Filter Units, 0.45um SFCA, 1000 mL; 12/Pk EW-06731-28 Mfr # 161-0045
COMPARE
1000 0.45 Yes
$452.00USD / Pkg of 12
Thermo Scientific Nalgene 291-4520 Bottletop Filters, 0.2um SFCA, 500 mL; 12/Pk EW-06731-30 Mfr # 291-4520
COMPARE
500 0.2 Yes
$219.00USD / Pkg of 12
Thermo Scientific Nalgene 291-4545 Bottletop Filters, 0.45um SFCA, 500 mL; 12/Pk EW-06731-32 Mfr # 291-4545
COMPARE
500 0.45 Yes
$219.00USD / Pkg of 12
Thermo Scientific Nalgene 292-4520 Bottletop Filters, 0.2um SFCA, 1000 mL; 12/Pk EW-06731-33 Mfr # 292-4520
COMPARE
1000 0.2 Yes
$319.00USD / Pkg of 12
Thermo Scientific Nalgene 290-4520 Bottletop Filters, 0.2um SFCA, 150 mL; 12/Pk EW-06731-34 Mfr # 290-4520
COMPARE
150 0.2 Yes
$159.00USD / Pkg of 12
Showing 1- 10 of 10
Need Help with this product?
Call Us At1-847-549-7600
Live chat