Thermo Scientific NanoDrop ND-3300 Fluorospectrometer
Saves time and space while utilizing as little as 1 µL of sample
  • Pipette your sample directly onto the optical surface — no need for separate cuvettes
  • Obtain readings in as little as 5 seconds!
  • Small footprint works gerat for even the most space-limited lab
The Thermo Scientific NanoDrop fluorospectrometer provides fluorescent analysis of 1 µL samples from 400 to 750 nm. Unit excites the sample from one of three LED sources; UV, blue, or white. The direct coupling of the sample to the optics and the proprietary signal processing for white LED applications enable measurements across a wide range of wavelengths — no need for filter changes.
The small sample size allows you to minimize loss, making units ideal for precious samples. Eliminating the need for cuvettes helps reduce error in results, giving you confidence that the results obtained are accurate.
The fluorospectrometer base measures only 5-1/2” x 7-7/8” (14.0 x 20.0 cm), taking up minimal lab space.
ITEM(S) ON SALE
$13,600.00 - $15,219.51USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bandwidth Max Wavelength (nm) Min Wavelength (nm)
Availability Pricing
Thermo Scientific ND-3300 Fluorospectrometer 83060-50 EW-83060-50 Mfr # ND-3300
COMPARE
750 400
$13,600.00USD / Each
Thermo Scientific ND3300LAPTOP Fluorospectrometer with Laptop EW-83061-56 Mfr # ND3300LAPTOP
COMPARE
280 260
$15,219.51USD / Each
$15,600.00USD / Each