Thermo Scientific Nunc Conical-bottom Centrifuge Tubes
Developed with your research needs in mind!
  • Reusable, collapsible and recyclable plastic rack is environmentally friendly
  • Large writing area provides ample space for labeling your samples
  • High RCF rating enables greater range of use and applications
These conical-bottom centrifuge tubes were developed for function and flexibility. The tubes provide a large writing space to label your samples. Polypropylene material is durable and can handle high centrifugation speeds, allowing for greater range of use in the lab. The plastic rack is collapsible and will break down when not in use, saving valuable lab space. The rack is manufactured with recyclable materials, lessening environmental impact and reducing waste in the lab.
$255.00 - $424.00USD / Case of 300
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Bottom Shape Tube Capacity (mL) Tube Material
Availability Pricing
Thermo Scientific Nunc 339650 Conical Centrifuge Tubes 500/CS, 15 mL EW-22999-80 Mfr # 339650
COMPARE
Conical 15 Polypropylene
$302.00USD / Case of 500
Thermo Scientific Nunc 339651 Conical Centrifuge Tubes 500/CS, RACKED 15 mL EW-22999-82 Mfr # 339651
COMPARE
Conical 15 Polypropylene
$304.00USD / Case of 500
Thermo Scientific Nunc 339652 Conical Centrifuge Tubes 500/CS, 50 mL EW-22999-84 Mfr # 339652
COMPARE
Conical 50 Polypropylene
$424.00USD / Case of 500
Thermo Scientific Nunc 339653 Conical Centrifuge Tubes 300/CS, RACKED 50 mL EW-22999-86 Mfr # 339653
COMPARE
Conical 50 Polypropylene
$255.00USD / Case of 300