Thermo Scientific™ Nunc™ Edge™ Sterile 96-Well Microplates
Minimize plate evaporation and the risks associated with edge effect
  • Excellent optical performance
  • Reduced edge effects
  • Use 38% more of your plate – cells are still viable
The Edge™ 2.0 96-well plates provide an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect. The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL of sterile water, buffer, or media. When filled with fluid, the surrounding moat acts as an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste. Choose from Nunclon™ treated or non-treated surfaces.
$413.00 - $500.00USD / Case of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Surface Treatment Well Volume (µL)
Availability Pricing
Thermo Scientific Nunc Edge™ Microplates with Lid, 96-Well, Polystyrene, Sterile, Nunclon™ Treated; 50/Cs EW-01930-64 Mfr # 167425
COMPARE
Polystyrene Nunclon™ Delta 400
$500.00USD / Case of 50
Thermo Scientific Nunc Edge™ Microplates with Lid, 96-Well, Polystyrene, Sterile, Non-Treated; 50/Cs EW-01930-66 Mfr # 267427
COMPARE
Polystyrene Non-Treated 400
$413.00USD / Case of 50