Thermo Scientific Nunc Roller Bottles
Boost gas exchange with increased overall growth area
  • Pleated surface increases growth area by over 40%
  • Prevent misthreading with easy on/off caps
Preserve your most precious samples by relying on these polystyrene (PS) roller bottles— bottles have been tested for reliability and reproducibility. Choose from an industry standard growth area of 850 cm2 or a pleated surface of 1450 cm2. The innovative pleated design provides over 40% additional growth area with the same external dimensions.
Easily view your culture growth via microscope through the viewing panels which run along the length of the bottle. Easy on/off caps (included) are engineered to prevent misthreading while providing a secure seal. Unique indentation at bottom lets you hold the bottle from the bottom, aiding in comfort and convenience when pouring your samples.
$202.00 - $292.00USD / Case of 20
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Closure Culture Area (cm²)
Availability Pricing
Thermo Scientific Nunc 182702 Roller Bottle; Smooth/Nonvented, 2/Pk, 20Pk/Cs EW-01836-11 Mfr # 182702
COMPARE
2000 Solid 850
$233.50USD / Case of 20
Thermo Scientific Nunc 182720 Roller Bottle; Smooth/Nonvented, 20/Pk 20Pk/Cs EW-01836-12 Mfr # 182720
COMPARE
2000 Solid 850
$202.00USD / Case of 20
Thermo Scientific Nunc 183902 EZ Roller Bottle; Smooth/Deep/Vent, 2/Pk 20Pk/Cs EW-01836-17 Mfr # 183902
COMPARE
2000 Vented 850
$292.00USD / Case of 20