Thermo Scientific Nunc Round-Bottom Tubes
PTFE tubes withstand RCF up to 30,000 x g in a close fitting holder. They are available with or without screw caps. Autoclavable. All other tubes withstand RCF up to 50,000 x g when used in a refrigerated centrifuge. All tubes without caps accept snap-on polypropylene closures. Autoclavable. Thermo Scientific Nalgene®.
ITEM(S) ON SALE
$22.90 - $150.20USD / Pkg of 10
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Bottom Shape Tube Capacity (mL) Tube Material
Availability Pricing
Thermo Scientific Nunc 3117-0120 Round-bottom tubes; 12 mL; PC; 10/pack
Buy More Save More
EW-06329-31 Mfr # 3117-0120
COMPARE
Round 12 Polycarbonate
$55.10USD / Pkg of 10
Thermo Scientific Nunc 3117-0150 Polycarbonate test tubes; 15 mL; 10/pack EW-06329-32 Mfr # 3117-0150
COMPARE
Round 15 Polycarbonate
$22.90USD / Pkg of 10
$57.25USD / Pkg of 10
Thermo Scientific Nunc 3117-0380 Round-bottom tubes; 38 mL; PC; 10/pack EW-06329-55 Mfr # 3117-0380
COMPARE
Round 38 Polycarbonate
$46.68USD / Pkg of 10
$58.35USD / Pkg of 10
Thermo Scientific Nunc 3117-0500 Polycarbonate Round-bottom tubes; 50 mL; 10/pack
Buy More Save More
EW-06329-60 Mfr # 3117-0500
COMPARE
Round 50 Polycarbonate
$64.30USD / Pkg of 10
Thermo Scientific Nunc 3117-9500 Round-bottom tubes; 50 mL; PC; 10/pack EW-06329-62 Mfr # 3117-9500
COMPARE
Round 50 Polycarbonate
$54.44USD / Pkg of 10
$68.05USD / Pkg of 10
Thermo Scientific Nunc 3117-1000 Round-bottom tubes; 100 mL; PC; 10/pack EW-06329-72 Mfr # 3117-1000
COMPARE
Round 100 Polycarbonate
$110.64USD / Pkg of 10
$138.30USD / Pkg of 10
Thermo Scientific Nunc 3110-0120 Round-bottom tubes; 12 mL; PPCO; 10/pack EW-06330-31 Mfr # 3110-0120
COMPARE
Round 12 Polypropylene
$51.52USD / Pkg of 10
$57.25USD / Pkg of 10
Thermo Scientific Nunc 3110-0150 Round-bottom tubes; 15 mL; PPCO; 10/pack EW-06330-32 Mfr # 3110-0150
COMPARE
Round 15 Polypropylene
$32.15USD / Pkg of 10
$64.30USD / Pkg of 10
Thermo Scientific Nunc 3110-0160 Round-bottom test tubes; 16 mL; PPCO; 10/pack EW-06330-40 Mfr # 3110-0160
COMPARE
Round 16 Polypropylene
$33.22USD / Pkg of 10
$66.45USD / Pkg of 10
Thermo Scientific Nunc 3110-0380 Round-bottom tubes; 38 mL; PPCO; 10/pack EW-06330-55 Mfr # 3110-0380
COMPARE
Round 38 Polypropylene
$32.15USD / Pkg of 10
$64.30USD / Pkg of 10
Thermo Scientific Nunc 3110-0500 Round-bottom test tubes; 50 mL; PPCO; 10/pack
Buy More Save More
EW-06330-60 Mfr # 3110-0500
COMPARE
Round 50 Polypropylene
$70.20USD / Pkg of 10
Thermo Scientific Nunc 3110-1000 Round-bottom tubes; 100 mL; PPCO; 10/pack
Buy More Save More
EW-06330-70 Mfr # 3110-1000
COMPARE
Round 100 Polypropylene
$150.20USD / Pkg of 10
Thermo Scientific Nunc 3110-9500 Round-bottom tubes; 50 mL; PPCO; 10/pack EW-06330-75 Mfr # 3110-9500
COMPARE
Round 50 Polypropylene
$42.95USD / Pkg of 10
$85.90USD / Pkg of 10
Showing 1- 13 of 13