Thermo Scientific™ Nunc™ MicroWell™ 96-Well Sterile Microplates with Nunclon™ Delta Surface Treatment
Certified Nunclon™ surface ensures optimum cell attachment and growth—use for scale up, cloning, viral titration, and fusion
  • Tested for cell growth and plating efficiency to ensure consistent biological performance
  • Raised well rims reduce risk of cross contamination
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Plates are optimized for robotics, plate readers and liquid handlers
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture microplates. These plates receive the proprietary Nunclon™ Delta surface treatment to ensure consistent cell growth. Lids feature condensation rings that reduce evaporation and eliminate contamination.
$399.00 - $401.00USD / Case of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sterile Well Shape Well Volume (mL)
Availability Pricing
Thermo Scientific Nunc MicroWell™ Cell Culture Microplates with Lid, 96-Well, Polystyrene, Nunclon™ Delta Treated, Round, 0.3 mL; 50/Cs EW-01930-10 Mfr # 163320
COMPARE
Yes Round 0.3
$399.00USD / Case of 50
Thermo Scientific Nunc MicroWell™ Cell Culture Microplates with Lid, 96-Well, Polystyrene, Nunclon™ Delta Treated, Flat, 0.4 mL; 50/Cs EW-01930-13 Mfr # 167008
COMPARE
Yes Flat 0.4
$401.00USD / Case of 50