Thermo Scientific™ Nunc™ EasYFlask™ Sterile Cell Culture Flasks with Nunclon™ Delta Surface Treatment
Nunclon™ Delta surface treatment provides optimal cell attachment and growth
  • EasYFlask™ ergonomic design features a 1/3 turn to open and close the cap
  • Virgin polystyrene provides excellent optical quality
  • Sterilized by gamma radiation
  • Large-bore necks—easy pipette and cell scraper access
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks. These flasks receive the proprietary Nunclon™ Delta surface treatment to ensure consistent cell growth. Low profile design maximizes incubator space. Flasks feature volume graduations printed and molded on both sides and slightly angled side walls allow for complete viewing under a microscope.
Vented caps feature a set vent position that produces a finger-felt positive stop when fully open for gas exchange and the Y on the cap provides a visual indication of the vent/close position when stacked in incubators. Filtered caps allow constant airflow and minimize chances of contamination.
$314.00 - $632.00USD / Case of 200
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Cap Type Culture Area (cm²) Working Volume (mL)
Availability Pricing
Thermo Scientific Nunc EasYFlask™ Cell Culture Flask, Polystyrene, Sterile, Treated, Angled Neck, Vent Cap, 25 cm2; 200/Cs EW-01930-48 Mfr # 156340
COMPARE
Vented 25 7
$546.00USD / Case of 200
Thermo Scientific Nunc EasYFlask™ Cell Culture Flask, Polystyrene, Sterile, Treated, Angled Neck, Vent Cap, 75 cm2; 100/ Cs EW-01930-49 Mfr # 156472
COMPARE
Vented 75 25
$522.00USD / Case of 100
Thermo Scientific Nunc EasYFlask™ Cell Culture Flask, Polystyrene, Sterile, Treated, Angled Neck, Filter Cap, 25 cm2; 200/Cs EW-01930-53 Mfr # 156367
COMPARE
Filter 25 7
$632.00USD / Case of 200
Thermo Scientific Nunc EasYFlask™ Cell Culture Flask, Polystyrene, Sterile, Treated, Angled Neck, Filter Cap, 75 cm2; 100/ Cs EW-01930-54 Mfr # 156499
COMPARE
Filter 75 25
$530.00USD / Case of 100
Thermo Scientific Nunc EasYFlask™ Cell Culture Flask, Polystyrene, Sterile, Treated, Angled Neck, Filter Cap, 175 cm2; 30/ Cs EW-01930-55 Mfr # 159910
COMPARE
Filter 175 55
$320.00USD / Case of 30
Thermo Scientific Nunc EasYFlask™ Cell Culture Flask, Polystyrene, Sterile, Treated, Angled Neck, Vent Cap, 175 cm2; 30/ Cs EW-01930-56 Mfr # 159920
COMPARE
Vented 175 55
$314.00USD / Case of 30