This product is now discontinued, View Related Products
Thermo Scientific Orion Star A100 Series Benchtop Dissolved Oxygen Meter
Make it easy and affordable to take simple, accurate dissolved oxygen measurements
  • Oxygen measurement directly in % saturation or mg/L (ppm)
  • Large LCD displays Conductivity along with temperature
  • Auto-Read™ function locks in stable reading
  • Nonvolatile memory holds up to 50 data points
  • AC power adapter or four AA batteries (sold separately)
Thermo Scientific Orion Star A112 benchtop dissolved oxygen and temperature meter combines simplicity with accuracy. Large informative display shows readings, battery life, electrode status, meter mode, power source and more. Features include automatic temperature, salinity and barometric pressure corrections. Simple button layout and onscreen messages help with calibration and setup menu choices.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Dissolved Oxygen Accuracy Min Dissolved Oxygen Probe Included
Availability Pricing
Thermo Scientific STAR A113 Star A113 Dissolved Oxygen Benchtop Meter EW-53101-21 Mfr # STARA1130
COMPARE
±0.5% of reading 0 to 20 ppm; 0 to 200% No
Call For Price
Thermo Scientific STAR A113 Star A113 Dissolved Oxygen Benchtop Meter Kit EW-53101-23 Mfr # STARA1135
COMPARE
±0.5% of reading 0 to 20 ppm; 0 to 200% Yes