Thermo Scientific Orion Star A200 pH/mV/Conductivity/TDS/Salinity/Resistivity/Temperature Benchtop Meter
Durability and dependability make it the ideal choice for every lab, every sample, everyday
  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Meets US Pharmacopoeia (USP) requirement
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp
  • Individual method passwords provide better security
  • Option of AC power or four AA batteries (sold separtely)
Thermo Scientific Orion Star A215 meter offers pH, mV, RmV, conductivity, salinity, TDS, resistivity and temperatur. Meter features an intuitive user interface with plain language prompts. A large, backlit graphic LCD shows measurements with temperature along with electrode status, time/date, sample ID, user ID, and calibration data.
GLP-related features include 2000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
Up to five point automatic pH calibration with USA/NIST and Din buffers, auto-buffer recognition, and recalibration feature to fix calibration errors.
Selectable cell constant for use with 2- or 4- band conductivity probes and selectable reference temperatures of 5, 10, 15, 20, 25°C ensure the most accrate conductivity measurement.
Available as meter only and as complete meter kit configurations.
$2,160.00 - $2,810.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Salinity Max TDS Description
Availability Pricing
Thermo Scientific STAR A215 Star A215 pH/Conductivity Benchtop Meter Only EW-58825-36 Mfr # STARA2150
COMPARE
80 ppt 200 ppt Star A215 pH/Conductivity Benchtop Meter Only
$2,160.00USD / Each
Thermo Scientific STAR A215 Star A215 pH/Conductivity Benchtop Meter Kit EW-58825-38 Mfr # STARA2155
COMPARE
80 ppt 200 ppt Star A215 pH/Conductivity Benchtop Meter Kit
$2,810.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat