Thermo Scientific™ SureSTART™ Certified Crimp Top Vials
Certified performance vials are quality tested to deliver the reproducibility you need
  • 2-mL size is ideal for chromatography applications
  • Decrease your risk of failed analysis and sample reruns
  • 100% traceable through the manufacturing process
  • For use with 11-mm crimp seals
Meet your certification and documentation needs with the Thermo Scientific™ SureSTART™ specification certified autosampler crimp top vials. The crown and neck of these glass vials are formed to precise tolerances for secure crimping and improved autosampler handling (caps sold separately. Vials feature write-on patches and have graduations at 0.5, 1.0, and 1.5 mL.
Choose from Level 2 or 3 compliance. Level 2 products have very low metal contents, extractables, and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. Level 3 products are packed in clean rooms to protect against environmental factors. They contain the lowest levels of metals, extractables, and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes.
$42.70 - $50.20USD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Level Tube Color Volume (mL)
Availability Pricing
Thermo Scientific SureSTART™ Certified Crimp Top Vial, Level 3, Clear Glass, 2 mL; 100/PK EW-98811-01 Mfr # 6PCV11-1P
COMPARE
3 Clear 2
$42.70USD / Pkg of 100
Thermo Scientific SureSTART™ Certified Crimp Top Vial, Level 2, Amber Glass, 2 mL; 100/PK EW-98811-02 Mfr # 6ACV11-2P
COMPARE
2 Amber 2
$50.20USD / Pkg of 100