Torque Wrenches
  • FEATURES
    • Dual scale, calibrated dual direction
    • Positive lock with spring loaded pull down lock ring
    • Fast, accurate and easy to set
    • Easy to read laser marked scales
    • Most feature Quick Release buttons which provide good socket retention as well as easy socket removal
    • Accuracy: ± 4% clockwise ± 6% CCW of indicated value, CW, from 20 to 100% of full scale
    • Meets or exceeds ASME B107.14M-1994, ISO 6789 standards
    • Individually serialized with matching certificate of calibration traceable to NIST
Micro-adjustable torque wrenches; metal handle English scale
The metal handle series offers the user a rugged, industrial strength torque wrench able to withstand the rigors of professional use. handles are lightly knurled for a tough, non-slip grip. There are 34 models of metal handle micrometer adjustable torque wrenches available
$327.50 - $415.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity
Availability Pricing
COMPARE
250 in lb / 27.7 N m
$327.50USD / Each
COMPARE
250 ft lb / 332 N m
$415.50USD / Each