Traceable® Calibrated Monitoring Hygrometers
Simultaneously displays time-of-day, temperature, and humidity
  • Available with a NIST-traceable or A2LA accredited (1750.01) calibration certificate depending on model
  • Features min/max memory, memory clear, °F/°C switchable, 12/24-hour clock
  • At the touch of a button, memory recalls the highest and lowest temperature and humidity readings
Clock displays AM/PM or 24-hour military time to the exact minute with 0.01% accuracy. Noting changes is easy with triple display. Internal sensors make it ideal for use in hoods, storerooms, clean rooms, incubators, drying chambers, and environmental cabinets. Temperature range is 32.0 to 122.0°F and 0.0 to 50.0°C. Resolution is 0.1° and accuracy is ±1°C. Relative humidity range is 20 to 90%. Resolution is 1% RH and accuracy is ±5% RH mid-range, otherwise 8%. At the touch of a button, memory recalls highest and lowest temperature and humidity readings. Permits monitoring conditions overnight, on weekends, or any time period.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$46.50 - $62.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RH Range (%) Max Temperature (° F) Description
Availability Pricing
COMPARE
90 122 NIST Calibrated Monitoring Hygrometer
$46.50USD / Each
COMPARE
90 122 A2LA-Accredited NIST Calibrated Monitoring Hygrometer
$62.50USD / Each