Traceable® Jumbo-Display Refrigerator/Freezer Thermometers with Calibration
Alarm provides alert when temperature rises above or falls below a set point
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory│Glass bottle probe are sealed and filled with unique nontoxic glycol solution
Thermometer is designed for monitoring freezers, water baths, heating blocks, incubators, refrigerators and other general-purpose applications. Display simultaneously shows maximum, minimum, and current probe temperatures. Min/max monitors high/low readings for any time period. Alarm is programmable in 1° increments. The 10-ft (3 m) micro-cable with solid-state probe permits doors to close on it.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2005(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
ITEM(S) ON SALE
$62.90 - $75.65USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Display Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
1..5" Display 122 -40
$75.65USD / Each
$89.00USD / Each
COMPARE
1..5" Display 122 -40
$71.40USD / Each
$84.00USD / Each
COMPARE
1..5" Display 122 -40
$62.90USD / Each
$74.00USD / Each