Traceable® Scientific Thermistor Thermometer Replacement Probe
Optimize performance of your thermometer
  • Probe is submersible up to the handle for ideal measuring
  • Features a temperature range of –58 to 302°F (–50 to 150°C)
  • Probe has 0.1 cm diameter, 7 cm stem length, 14 cm overall length, and 152 cm cable length
Compatible for use with the Traceable Scientific Thermistor Thermometer 90080-12, meter sold separately.
$76.50 - $126.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Penetration Probe for Scientific Thermistor Thermometer 90080-12
$76.50USD / Each
COMPARE
Micro Probe for Scientic Thermistor Thermometer 90080-12
$126.00USD / Each