Traceable® Thermohygrometers with RS-232 and Calibration
Accurately measure temperature and humidity
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Microprocessor-based for quick and easy calibration
Simultaneously displays both temperature and relative humidity on the dual 1-1/8-inch LCD display. These meters feature data hold, MIN/MAX, memory storage, selectable °F/°C, low-battery indicator, and auto-off. Model 4385 (37950-02) also calculates dew point and features an alarm for out-of-range conditions.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$200.00 - $402.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Battery Max RH Range (%) Max Temperature (° F)
Availability Pricing
COMPARE
9V battery (1) 95 122
$200.00USD / Each
COMPARE
9V battery (1) 95 122
$402.00USD / Each