Traceable® Waterproof Remote Probe Digital Thermometer with Calibration
Water resistant and shockproof—ideal for any area!
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Display updates every second
  • Features MIN, MAX, and selectable °F/°C
Water-resistant remote probe thermometer is perfect for wet areas—case is IP61 rated and shockproof, and probe is impervious to acid and solvents. Probe with 10-ft (3-m) flexible cord makes it easy to take temperature measurements in hard to reach areas. Supplied with suction cups, magnet, and hook-and-loop tabs to attach to any surface.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2005(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
ITEM(S) ON SALE
$47.17 - $53.55USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
COMPARE
572 -58 Traceable® Water-Resistant Remote Probe Thermometer with Calibration; ±1°C accuracy (-20 to 100°C)
$47.17USD / Each
$55.50USD / Each
COMPARE
572 -58 Traceable® Water-Resistant Remote Probe Thermometer Ultra with Calibration; ±0.5°C accuracy at tested points
$53.55USD / Each
$63.00USD / Each